Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 1 - 2 | 434 | 78 | |
GenBank mRNA | AK032814.1 | - | 1 - 2 | 372 | 78 | |
AK048355.1 | - | 1 - 2 | 396 | 78 | ||
AK053857.1 | - | 1 - 2 | 424 | 78 | ||
AK085211.1 | - | 1 - 2 | 425 | 78 | ||
AK165945.1 | - | 1 - 2 | 401 | 78 | ||
AY557610.1 | - | 1 - 2 | 425 | 78 | ||
BC038279.1 | - | 1 - 2 | 442 | 78 | ||
BC141180.1 | - | 1 - 2 | 92 | 9055792 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
