Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 4 - 5 | 2561 | 105 | |
GenBank mRNA | AK048355.1 | - | 4 - 5 | 2523 | 105 | |
AK085211.1 | - | 4 - 5 | 2552 | 105 | ||
AK165945.1 | - | 4 - 5 | 2528 | 105 | ||
AY557610.1 | - | 4 - 5 | 2552 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
