Assay Details
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 7 - 8 | 2955 | 67 | |
GenBank mRNA | AY557610.1 | - | 7 - 8 | 2946 | 67 | |
BC046463.1 | - | 3 - 4 | 252 | 67 | ||
BC141180.1 | - | 6 - 7 | 2451 | 9055792 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
