Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039371.2 | NP_001034460.2 | 1 - 2 | 425 | 80 | |
GenBank mRNA | BB615842.1 | - | 1 - 2 | 418 | 80 | |
BC094925.1 | - | 1 - 2 | 351 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
