Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039371.2 | NP_001034460.2 | 5 - 6 | 1070 | 72 | |
GenBank mRNA | AK029695.1 | - | 2 - 3 | 228 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK029695.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
