Assay Details
Gene Symbol: Npepps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008942.2 | NP_032968.2 | 9 - 10 | 1218 | 73 | |
GenBank mRNA | AK133898.1 | - | 9 - 10 | 1026 | 73 | |
BC009653.1 | - | 9 - 10 | 1157 | 3487224 | 73 | |
BC086798.1 | - | 9 - 10 | 1210 | 6812900 | 73 | |
BC098212.1 | - | 9 - 10 | 1218 | 6825181 | 73 | |
U35646.1 | - | 9 - 10 | 1205 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
