Assay Details
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271472.1 | NP_001258401.1 | 3 - 4 | 476 | 75 | |
NM_001271473.1 | NP_001258402.1 | 3 - 4 | 476 | 75 | ||
NM_022723.3 | NP_073560.2 | 3 - 4 | 476 | 75 | ||
GenBank mRNA | AF276425.1 | - | 3 - 4 | 343 | 75 | |
AK041590.1 | - | 3 - 4 | 459 | 75 | ||
BC066066.1 | - | 3 - 4 | 434 | 6850999 | 75 | |
BC080278.1 | - | 3 - 4 | 444 | 30354974 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
