Assay Details
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207225.1 | NP_997108.1 | 22 - 23 | 2939 | 71 | |
GenBank mRNA | AK029933.1 | - | 19 - 20 | 2491 | 71 | |
AK155250.1 | - | 23 - 24 | 3125 | 71 | ||
BC066052.1 | - | 22 - 23 | 2939 | 6827645 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
