Assay Details
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134252.3 | NP_599013.1 | 9 - 10 | 1126 | 73 | |
GenBank mRNA | AF481480.2 | - | 1395 | 73 | ||
AY095352.1 | - | 7 - 8 | 960 | 73 | ||
BC117934.1 | - | 9 - 10 | 1126 | 40097778 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
