Assay Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021530.2 | NP_067505.2 | 14 - 15 | 2081 | 94 | |
GenBank mRNA | AF224508.1 | - | 14 - 15 | 2027 | 94 | |
AK034237.1 | - | 14 - 15 | 2083 | 94 | ||
AK047863.1 | - | 7 - 8 | 1015 | 94 | ||
AK171610.1 | - | 13 - 14 | 1857 | 94 | ||
AK173014.1 | - | 14 - 15 | 1915 | 94 | ||
BC030388.1 | - | 14 - 15 | 2043 | 5401143 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
