Assay Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021530.2 | NP_067505.2 | 16 - 17 | 66 | ||
GenBank mRNA | AF224508.1 | - | 16 - 17 | 66 | ||
AK034237.1 | - | 16 - 17 | 66 | |||
AK039404.1 | - | 16 - 17 | 66 | |||
AK047863.1 | - | 9 - 10 | 66 | |||
AK171610.1 | - | 15 - 16 | 66 | |||
AK173014.1 | - | 16 - 17 | 66 | |||
BC030388.1 | - | 16 - 17 | 5401143 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
