Assay Details
Gene Symbol: Npw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099664.1 | NP_001093134.1 | 1 - 2 | 429 | 63 | |
GenBank mRNA | AK131881.1 | - | 1 - 2 | 211 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124871 | NM_001099664.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124871 | AK131881.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124871 | NM_001099664.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124871 | AK131881.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13284758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13241627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13200826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13512072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098836 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13338011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13200830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31098829 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10448691 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448692 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448693 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10448690 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700014639.mm.1 | TC1700001635.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700014639.mm.1 | NM_001099664.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700014639.mm.1 | NM_001099664.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700014640.mm.1 | TC1700001635.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700014640.mm.1 | NM_001099664.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700014640.mm.1 | NM_001099664.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258556 | - | - |
