Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK209503.1 | - | 1 - 2 | 138 | 79 | |
AK220149.1 | - | 2 - 3 | 153 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
