Assay Details
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163691.1 | NP_001157163.1 | 21 - 22 | 4587 | 108 | |
NM_021415.4 | NP_067390.4 | 21 - 22 | 4587 | 108 | ||
GenBank mRNA | AK122452.1 | - | 10 - 11 | 1560 | 108 | |
AK135951.1 | - | 3 - 4 | 334 | 108 | ||
AK147242.1 | - | 21 - 22 | 4588 | 108 | ||
BC138026.1 | - | 21 - 22 | 4426 | 8861045 | 108 | |
CA327685.1 | - | 4 - 5 | 408 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
