Assay Details
Gene Symbol: Chtf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145409.2 | NP_663384.2 | 13 - 14 | 1730 | 73 | |
GenBank mRNA | AK052673.1 | - | 12 - 13 | 1941 | 73 | |
AK077947.1 | - | 13 - 14 | 1697 | 73 | ||
BC024142.1 | - | 13 - 14 | 1717 | 5149957 | 73 | |
BC083096.1 | - | 10 - 11 | 2448 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
