Assay Details
Gene Symbol: Nrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013724.2 | NP_038752.2 | 17 - 18 | 2919 | 132 | |
GenBank mRNA | AB020741.1 | - | 17 - 18 | 2844 | 132 | |
AB035267.1 | - | 17 - 18 | 2597 | 132 | ||
AK135285.1 | - | 2 - 3 | 137 | 132 | ||
BC139029.1 | - | 17 - 18 | 2686 | 8862051 | 132 | |
BC139031.1 | - | 17 - 18 | 2686 | 8862053 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
