Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025779.1 | NP_001020950.1 | 5 - 6 | 950 | 68 | |
NM_011799.2 | NP_035929.1 | 5 - 6 | 1026 | 68 | ||
GenBank mRNA | AJ009559.1 | - | 5 - 6 | 1043 | 68 | |
AJ223087.1 | - | 5 - 6 | 1028 | 68 | ||
AK012192.1 | - | 5 - 6 | 980 | 68 | ||
AK085020.1 | - | 5 - 6 | 980 | 68 | ||
AK162669.1 | - | 5 - 6 | 988 | 68 | ||
AK166003.1 | - | 5 - 6 | 1031 | 68 | ||
BC052434.1 | - | 5 - 6 | 950 | 6837113 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
