Assay Details
Gene Symbol: Bcl2l15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142959.1 | NP_001136431.1 | 3 - 4 | 622 | 106 | |
GenBank mRNA | BC027668.1 | - | 3 - 4 | 458 | 106 | |
BC117780.1 | - | 3 - 4 | 622 | 40091931 | 106 | |
BC117781.1 | - | 3 - 4 | 622 | 40091934 | 106 | |
BC128505.1 | - | 3 - 4 | 622 | 40091935 | 106 | |
BC128506.1 | - | 3 - 4 | 622 | 40091936 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975874 | BC027668.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975874 | NM_001142959.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975877 | AV381247.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975877 | NM_001142960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975873 | BC052523.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975873 | NM_001142959.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975873 | NM_001142960.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477943 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942287 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2944 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430526 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680703 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577925 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867833 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82583 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332809 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1548 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739921 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67718 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13581 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121222 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145715 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975874 | BC027668.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975874 | NM_001142959.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975873 | BC052523.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975873 | NM_001142959.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975873 | NM_001142960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975877 | AV381247.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975877 | NM_001142960.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145715 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02625426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02603983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02889547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02901259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02603982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22388315 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10494976 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494974 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494977 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494975 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452614_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10494972 | - | - |
Mouse Genome 430 2.0 Array | 1452614_at | - | - |
Mouse Genome 430 2.0 Array | 1452614_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229672_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229672_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300008597.mm.1 | TC0300001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008597.mm.1 | NM_001142959.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008597.mm.1 | NM_001142959.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008598.mm.1 | TC0300001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008598.mm.1 | NM_001142959.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008598.mm.1 | NM_001142959.1 | Refseq |
Mouse Expression Set 430 | 1452614_at | - | - |
Mouse Expression Set 430 | 1452614_PM_at | - | - |
Murine Genome U74v2 Set | 168860_PM_i_at | - | - |
Murine Genome U74v2 Set | 168860_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1452614_at | - | - |
Mouse Genome 430A 2.0 Array | 1452614_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452614_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452614_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257368 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257368 | - | - |
