Assay Details
Gene Symbol: Arhgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152801.2 | NP_690014.2 | 3 - 4 | 568 | 109 | |
GenBank mRNA | AF393831.1 | - | 3 - 4 | 568 | 109 | |
AK028642.1 | - | 2 - 3 | 583 | 109 | ||
AK040468.1 | - | 3 - 4 | 390 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
