Assay Details
Gene Symbol: Rad51d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277938.1 | NP_001264867.1 | 8 - 9 | 101 | ||
NM_001277939.1 | NP_001264868.1 | 8 - 9 | 101 | |||
NM_001277941.1 | NP_001264870.1 | 7 - 8 | 101 | |||
NM_011235.4 | NP_035365.1 | 8 - 9 | 101 | |||
NR_102718.1 | 8 - 9 | 101 | ||||
NR_102719.1 | 7 - 8 | 101 | ||||
NR_102720.1 | 7 - 8 | 101 | ||||
GenBank mRNA | AB007040.1 | - | 8 - 9 | 101 | ||
AB052829.1 | - | 8 - 9 | 897 | 101 | ||
AB052831.1 | - | 7 - 8 | 793 | 101 | ||
AB052832.1 | - | 7 - 8 | 778 | 101 | ||
AB052833.1 | - | 7 - 8 | 777 | 101 | ||
AB052834.1 | - | 8 - 9 | 935 | 101 | ||
AF034955.1 | - | 8 - 9 | 101 | |||
AK017835.1 | - | 8 - 9 | 101 | |||
AK036272.1 | - | 7 - 8 | 101 | |||
AK138553.1 | - | 8 - 9 | 101 | |||
AK147759.1 | - | 8 - 9 | 101 | |||
BC049136.1 | - | 8 - 9 | 6832014 | 101 | ||
CF746344.1 | - | 4 - 5 | 101 | |||
EU627688.1 | - | 8 - 9 | 101 | |||
Y15570.1 | - | 8 - 9 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
