Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025779.1 | NP_001020950.1 | 1 - 2 | 94 | 84 | |
GenBank mRNA | AK012192.1 | - | 1 - 2 | 124 | 84 | |
AK075975.1 | - | 1 - 2 | 124 | 84 | ||
AK085020.1 | - | 1 - 2 | 124 | 84 | ||
AK162669.1 | - | 1 - 2 | 135 | 84 | ||
AK166003.1 | - | 1 - 2 | 175 | 84 | ||
BC052434.1 | - | 1 - 2 | 94 | 6837113 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK012192.1, AK085020.1, AK166003.1, BC052434.1, NM_001025779.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
