Assay Details
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027081.3 | NP_081357.2 | 8 - 9 | 779 | 74 | |
GenBank mRNA | AK154949.1 | - | 8 - 9 | 801 | 74 | |
BC017544.1 | - | 3 - 4 | 212 | 74 | ||
BC030395.1 | - | 9 - 10 | 936 | 74 | ||
BC058958.1 | - | 4 - 5 | 338 | 74 | ||
BC080797.1 | - | 5 - 6 | 397 | 74 | ||
BC139003.1 | - | 8 - 9 | 753 | 8862025 | 74 | |
BQ444357.1 | - | 4 - 5 | 337 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
