Assay Details
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144900.2 | NP_659149.1 | 7 - 8 | 1049 | 62 | |
GenBank mRNA | AK088000.1 | - | 7 - 8 | 1010 | 62 | |
AK137001.1 | - | 7 - 8 | 1008 | 62 | ||
AK159279.1 | - | 7 - 8 | 1006 | 62 | ||
AK159306.1 | - | 7 - 8 | 1004 | 62 | ||
BC010319.1 | - | 5 - 6 | 634 | 62 | ||
BC021496.1 | - | 7 - 8 | 999 | 5346159 | 62 | |
BC023794.1 | - | 8 - 9 | 944 | 5351943 | 62 | |
BC023964.1 | - | 7 - 8 | 989 | 62 | ||
BC025618.1 | - | 7 - 8 | 1008 | 5320976 | 62 | |
BC025627.1 | - | 7 - 8 | 999 | 5320513 | 62 | |
BC025811.1 | - | 7 - 8 | 986 | 5096872 | 62 | |
BC032187.1 | - | 7 - 8 | 1049 | 5321268 | 62 | |
BC033435.1 | - | 7 - 8 | 945 | 4950746 | 62 | |
BC033471.1 | - | 7 - 8 | 989 | 4954119 | 62 | |
BC034177.1 | - | 6 - 7 | 754 | 62 | ||
BC035352.1 | - | 6 - 7 | 719 | 62 | ||
BC042435.1 | - | 7 - 8 | 999 | 4951288 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
