Assay Details
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028011.2 | NP_082287.1 | 3 - 4 | 263 | 105 | |
GenBank mRNA | AF395837.1 | - | 3 - 4 | 227 | 105 | |
AJ505007.1 | - | 3 - 4 | 224 | 105 | ||
AK009826.1 | - | 3 - 4 | 226 | 105 | ||
AK033712.1 | - | 3 - 4 | 265 | 105 | ||
AK083630.1 | - | 225 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK033712.1, NM_028011.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
