Assay Details
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162996.1 | NP_001156468.1 | 4 - 5 | 360 | 88 | |
NM_028153.1 | NP_082429.1 | 4 - 5 | 417 | 88 | ||
GenBank mRNA | AK005562.1 | - | 4 - 5 | 383 | 88 | |
BC055476.1 | - | 4 - 5 | 360 | 4218133 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
