Assay Details
Gene Symbol: Gm30339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035352.1 | - | 2 - 3 | 1617 | 150 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prickle homolog 1 (Drosophila) | Prickle1 |
1110058P22Rik, AW215793, Prickle, b2b019Clo, mpk1 |
-140.3kb | Anti-sense | Chr. 15 - 93499111 - 93596266 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 9430014N10 gene | 9430014N10Rik | 166.4kb | Anti-sense | Chr. 15 - 93904856 - 93927111 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
