Assay Details
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175229.3 | NP_780438.2 | 10 - 11 | 7572 | 82 | |
GenBank mRNA | AK090123.1 | - | 10 - 11 | 7533 | 82 | |
AK122248.1 | - | 1 - 2 | 4757 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
