Assay Details
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175229.3 | NP_780438.2 | 6 - 7 | 655 | 77 | |
GenBank mRNA | AK090123.1 | - | 6 - 7 | 616 | 77 | |
AK171001.1 | - | 7 - 8 | 928 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
