Assay Details
Gene Symbol: Thap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006173.1 | - | 2 - 3 | 486 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | WD repeat domain 62 | Wdr62 | 2310038K02Rik | -181.0 | Anti-sense | Chr. 7 - 30240138 - 30280427 on NCBI Build GRCm38 |
Downstream | CAP-GLY domain containing linker protein 3 | Clip3 |
1500005P14Rik, AI844915, Clipr-59 |
2.0kb | Sense | Chr. 7 - 30291688 - 30308367 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
