Assay Details
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011882.2 | NP_036012.1 | 3 - 4 | 482 | 93 | |
GenBank mRNA | AK054205.1 | - | 1 - 2 | 977 | 93 | |
AK140815.1 | - | 3 - 4 | 483 | 93 | ||
BC003873.1 | - | 3 - 4 | 291 | 93 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK054205.1, BC003873.1, NM_011882.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
