Assay Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 10 - 11 | 1593 | 60 | |
GenBank mRNA | AK035870.1 | - | 10 - 11 | 1594 | 60 | |
AK043769.1 | - | 10 - 11 | 1532 | 60 | ||
AK137169.1 | - | 10 - 11 | 1358 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
