Assay Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 21 - 22 | 3202 | 172 | |
GenBank mRNA | AK035870.1 | - | 21 - 22 | 3203 | 172 | |
AK040830.1 | - | 12 - 13 | 1626 | 172 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
