Assay Details
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 25 - 26 | 3612 | 105 | |
GenBank mRNA | AK035870.1 | - | 25 - 26 | 3613 | 105 | |
AK040830.1 | - | 16 - 17 | 2036 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
