Assay Details
Gene Symbol: Ikzf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011771.1 | NP_035901.1 | 5 - 6 | 872 | 62 | |
GenBank mRNA | AF001293.1 | - | 4 - 5 | 591 | 62 | |
AK004152.1 | - | 1 - 2 | 171 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
