Assay Details
Gene Symbol: Grb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010346.2 | NP_034476.1 | 12 - 13 | 1698 | 80 | |
GenBank mRNA | AK057640.1 | - | 12 - 13 | 1691 | 80 | |
AK145996.1 | - | 12 - 13 | 1514 | 80 | ||
AK168438.1 | - | 12 - 13 | 1482 | 80 | ||
AK220277.1 | - | 11 - 12 | 1589 | 80 | ||
BC003295.1 | - | 12 - 13 | 1441 | 3583896 | 80 | |
BC081547.1 | - | 11 - 12 | 1687 | 80 | ||
M94450.1 | - | 12 - 13 | 1720 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
