Assay Details
Gene Symbol: Cdk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109626.1 | NP_001103096.1 | 7 - 8 | 2718 | 74 | |
NM_001109628.1 | NP_001103098.1 | 7 - 8 | 2718 | 74 | ||
NM_026952.2 | NP_081228.2 | 7 - 8 | 2718 | 74 | ||
GenBank mRNA | AK049376.1 | - | 7 - 8 | 2364 | 74 | |
AK129237.1 | - | 7 - 8 | 1857 | 74 | ||
AK137734.1 | - | 5 - 6 | 588 | 74 | ||
AY072295.1 | - | 7 - 8 | 2709 | 74 | ||
BC116645.1 | - | 7 - 8 | 2701 | 40086381 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
