Assay Details
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038010.2 | NP_001033099.1 | 10 - 11 | 1676 | 84 | |
NM_020004.5 | NP_064388.2 | 10 - 11 | 1679 | 84 | ||
GenBank mRNA | AF254441.1 | - | 10 - 11 | 1616 | 84 | |
AK137358.1 | - | 10 - 11 | 1656 | 84 | ||
AK147042.1 | - | 10 - 11 | 1624 | 84 | ||
AK150225.1 | - | 10 - 11 | 1657 | 84 | ||
AK154866.1 | - | 10 - 11 | 1667 | 84 | ||
AK155347.1 | - | 10 - 11 | 1853 | 84 | ||
AK155791.1 | - | 10 - 11 | 1678 | 84 | ||
AK155842.1 | - | 10 - 11 | 1677 | 84 | ||
AK158079.1 | - | 10 - 11 | 1667 | 84 | ||
BC051952.1 | - | 10 - 11 | 1661 | 84 | ||
BC060050.1 | - | 10 - 11 | 1661 | 84 | ||
BC063752.1 | - | 10 - 11 | 1661 | 6333090 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
