Assay Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291440.1 | NP_001278369.1 | 4 - 5 | 923 | 80 | |
NM_053269.3 | NP_444499.1 | 4 - 5 | 755 | 80 | ||
GenBank mRNA | AK136204.1 | - | 4 - 5 | 769 | 80 | |
BC090648.1 | - | 699 | 30640414 | 80 | ||
BC141034.1 | - | 4 - 5 | 750 | 9055646 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
