Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 16 - 17 | 1563 | 64 | |
GenBank mRNA | AK004652.1 | - | 12 - 13 | 1215 | 64 | |
AK082323.1 | - | 9 - 10 | 816 | 64 | ||
AK082761.1 | - | 16 - 17 | 1564 | 64 | ||
AK141692.1 | - | 16 - 17 | 1582 | 64 | ||
AK145969.1 | - | 16 - 17 | 1572 | 64 | ||
BC034517.1 | - | 15 - 16 | 1499 | 4458839 | 64 | |
U97572.1 | - | 15 - 16 | 1547 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
