Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 20 - 21 | 1920 | 83 | |
GenBank mRNA | AK004652.1 | - | 16 - 17 | 1572 | 83 | |
AK082323.1 | - | 13 - 14 | 1174 | 83 | ||
AK082761.1 | - | 20 - 21 | 1921 | 83 | ||
BC034517.1 | - | 19 - 20 | 1856 | 4458839 | 83 | |
U97572.1 | - | 19 - 20 | 1904 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
