Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 4 - 5 | 262 | 98 | |
GenBank mRNA | AK082761.1 | - | 4 - 5 | 263 | 98 | |
AK141692.1 | - | 4 - 5 | 281 | 98 | ||
AK145969.1 | - | 4 - 5 | 271 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
