Assay Details
Gene Symbol: Dusp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026725.2 | NP_081001.1 | 1 - 2 | 362 | 70 | |
GenBank mRNA | AA048085.1 | - | 1 - 2 | 362 | 70 | |
AB183013.1 | - | 1 - 2 | 356 | 70 | ||
AK004912.1 | - | 1 - 2 | 253 | 70 | ||
BC069187.1 | - | 1 - 2 | 356 | 850475 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834310 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834309 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834314 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834313 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834311 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030638 | NM_026725.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905553 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165936 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030638 | NM_026725.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P260187 | NM_026725.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P260187 | AK004912.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834314 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834303 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834305 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834306 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834308 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834330 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834313 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834310 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834301 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834319 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834320 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20834296 | - | - |
Whole Mouse Genome G4122A | A_52_P260187 | NM_026725.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P260187 | AK004912.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01003074 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01003062 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01003065 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01003063 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834314 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834311 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834310 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834313 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20834309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20834301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00378523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01003075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01117328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01051782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834279 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00540071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00108358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01045217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00476489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00513023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00067776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00513017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01010147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01003062 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01051790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20834259 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10360322 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360323 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453074_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10360321 | - | - |
Mouse Genome 430 2.0 Array | 1453074_at | - | - |
Mouse Genome 430 2.0 Array | 1453074_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68440_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68440_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100029381.mm.1 | TC0100003577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029381.mm.1 | NM_026725.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029381.mm.1 | NM_026725.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029380.mm.1 | TC0100003577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029380.mm.1 | NM_026725.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029380.mm.1 | NM_026725.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029382.mm.1 | TC0100003577.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029382.mm.1 | NM_026725.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029382.mm.1 | NM_026725.2 | Refseq |
Mouse Expression Set 430 | 1453074_at | - | - |
Mouse Expression Set 430 | 1453074_PM_at | - | - |
Murine Genome U74v2 Set | 135181_f_at | - | - |
Murine Genome U74v2 Set | 135181_PM_f_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6100632 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160864 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160864 | - | - |
mouse-6 v1.1 (Illumina) | GI_55926214-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3160863 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160863 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_55926214-S | - | - |
