Assay Details
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205343.1 | NP_001192272.1 | 9 - 10 | 932 | 148 | |
NM_130887.3 | NP_570957.2 | 9 - 10 | 932 | 148 | ||
GenBank mRNA | AK053198.1 | - | 9 - 10 | 933 | 148 | |
AK086767.1 | - | 9 - 10 | 913 | 148 | ||
AK138980.1 | - | 9 - 10 | 905 | 148 | ||
BC132475.1 | - | 9 - 10 | 872 | 40130752 | 148 | |
BC137854.1 | - | 9 - 10 | 872 | 8860873 | 148 | |
BC144975.1 | - | 9 - 10 | 872 | 9053523 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
