Assay Details
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011019.3 | NP_035149.2 | 6 - 7 | 948 | 102 | |
GenBank mRNA | AB015978.1 | - | 6 - 7 | 1602 | 102 | |
AF058805.1 | - | 6 - 7 | 918 | 102 | ||
AK157508.1 | - | 1 - 2 | 118 | 102 | ||
BC137703.1 | - | 5 - 6 | 843 | 8860722 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
