Assay Details
Gene Symbol: Ylpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178363.3 | NP_848140.3 | 17 - 18 | 6258 | 125 | |
GenBank mRNA | CA980613.1 | - | 5 - 6 | 481 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
