Assay Details
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199198.2 | NP_954668.2 | 12 - 13 | 1326 | 66 | |
NR_028447.1 | 12 - 13 | 1337 | 66 | |||
NR_028448.1 | 11 - 12 | 1257 | 66 | |||
NR_028449.1 | 12 - 13 | 1337 | 66 | |||
GenBank mRNA | AK040235.1 | - | 12 - 13 | 1289 | 66 | |
AK045028.1 | - | 11 - 12 | 1537 | 66 | ||
AK154045.1 | - | 11 - 12 | 1221 | 66 | ||
BC013700.1 | - | 12 - 13 | 1290 | 66 | ||
BC064018.1 | - | 12 - 13 | 1253 | 3980699 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
