Assay Details
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199198.2 | NP_954668.2 | 7 - 8 | 942 | 68 | |
NR_028447.1 | 7 - 8 | 942 | 68 | |||
NR_028448.1 | 6 - 7 | 873 | 68 | |||
NR_028449.1 | 7 - 8 | 942 | 68 | |||
GenBank mRNA | AK040235.1 | - | 7 - 8 | 894 | 68 | |
AK045028.1 | - | 6 - 7 | 1153 | 68 | ||
AK154045.1 | - | 6 - 7 | 837 | 68 | ||
BC013700.1 | - | 7 - 8 | 895 | 68 | ||
BC064018.1 | - | 7 - 8 | 869 | 3980699 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
