Assay Details
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153171.4 | NP_694811.1 | 3 - 4 | 242 | 94 | |
GenBank mRNA | AF498319.1 | - | 3 - 4 | 291 | 94 | |
AK043268.1 | - | 3 - 4 | 242 | 94 | ||
BC049624.1 | - | 3 - 4 | 160 | 6585574 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
