Assay Details
Gene Symbol: Bcl2l15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142960.1 | NP_001136432.1 | 3 - 4 | 615 | 191 | |
GenBank mRNA | BC052523.1 | - | 3 - 4 | 550 | 191 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
