Assay Details
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 17 - 18 | 2252 | 108 | |
GenBank mRNA | BC055377.1 | - | 17 - 18 | 2174 | 30077916 | 108 |
M90388.1 | - | 17 - 18 | 2237 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
